Learn More
Novus Biologicals™ Mouse Galectin-12 ELISA Kit (Colorimetric)
Mouse Galectin-12 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276719
Catalog No. NBP276719
Add to cart
Description
Description
The Mouse Galectin-12 ELISA Kit (Colorimetric) measures mouse Galectin-12 in serum, plasma and other biological fluids.
Specifications
Specifications
31.25 to 2000 pg/mL
ELISA Kit (Colorimetric)
Storage is content dependent.
LGALS12
Biological Fluids, Plasma, Serum
85329
18.75 pg/mL
Mouse
1 Kit