Learn More
Abnova™ Mouse Gzmk (O35205) Full length Recombinant Protein
Used for SDS-PAGE
Manufacturer:  Abnova™ P6173
Catalog No. 89-946-931
Add to cart
Specifications
Specifications
O35205
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
30kDa
Escherichia coli expression system
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gzmk
Mouse ;Mouse
Yes
Escherichia coli expression system
Liquid
14945
Gzmk (Mouse) Recombinant Protein
100 ug
RUO
Gzmk
Escherichia coli
None