Learn More
Novus Biologicals™ Mouse HB-EGF ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP262763
Catalog No. NBP262763
Add to Cart
Description
Description
The Mouse HB-EGF ELISA Kit (Colorimetric) measures mouse HB-EGF in serum, plasma and other biological fluids.
Specifications
Specifications
15.625--1000 pg/mL
This kit recognizes Mouse HB-EGF in samples. No significant cross-reactivity or interference between Mouse HB-EGF and analogues was observed.
Biological Fluids, Plasma, Serum
1839
Mouse
Storage of components varies. See protocol for specific instructions.
9.375 pg/mL
ELISA Kit (Colorimetric)
ELISA
HBEGF
1 Kit
HB-EGF
Recommendations
Recommendations