Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse HBeAg Antibody ELISA Kit (Colorimetric)

Detection for Hepatitis B antibody

$732.50

Specifications

Species Mouse
Sample Type Serum, plasma
Research Discipline Cell Biology
Storage Requirements Store the unopened product at 2° to 8°C Protect from light. Do not use past expiration date.
Target HBeAg Antibody
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600311KT View Documents Novus Biologicals™
NBP2600311KIT
1 Kit Each for $732.50
Add to cart
 
Description

Description

The Mouse HBeAg Antibody ELISA Kit (Colorimetric) measures mouse HBeAg Antibody in serum, plasma.
Specifications

Specifications

Mouse
Cell Biology
HBeAg Antibody
HBeAg
Qualitative
Serum, plasma
Store the unopened product at 2° to 8°C Protect from light. Do not use past expiration date.
ELISA
ELISA Kit (Colorimetric)
50 to 100μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit