Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse HBsAg ELISA Kit (Colorimetric)

Direct ELISA kit for Hepatitis B antibody detection.

$732.50

Specifications

Species Mouse
Sample Type Serum, plasma
Research Discipline Cell Biology
Synonym HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S
Storage Requirements Store the unopened product at 2° to 8°C Protect from light. Do not use past expiration date.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB599681KT View Documents Novus Biologicals™
NBP2599681KIT
1 Kit Each for $732.50
Add to cart
 
Description

Description

The Mouse HBsAg ELISA Kit (Colorimetric) measures mouse HBsAg in serum, plasma.
Specifications

Specifications

Mouse
Cell Biology
Store the unopened product at 2° to 8°C Protect from light. Do not use past expiration date.
ELISA
HBVgp2, MBL2, S
Qualitative
Serum, plasma
HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S
HBsAg
944569
ELISA Kit (Colorimetric)
50 to 100μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit