Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse Hc (NP_034536, 77 amino acids) Partial Recombinant Protein

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P4491

Catalog No. 89-947-138


Add to cart

Specifications

Specifications

NP_034536
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
9kDa
Escherichia coli expression system
20 ug
RUO
C5|C5a|He
Hc
Escherichia coli
None
Escherichia coli expression system
Lyophilized
15139
Hc (Mouse) Recombinant Protein
≥95% by SDS-PAGE
Store at -20°C on dry atmosphere. After reconstitution with ice-cold sterilized water containing 2.5 mg/mL BSA, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
Hc
The ED50 1-3nm as determined by enzyme release assay of myeloperoxidase.
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit