Learn More
Novus Biologicals™ Mouse HSF1 ELISA Kit (Colorimetric)
Mouse HSF1 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276589
Catalog No. NBP276589
Add to cart
Description
Description
The Mouse HSF1 ELISA Kit (Colorimetric) measures mouse HSF1 in serum, plasma and other biological fluids.
Specifications
Specifications
0.781 to 50 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Mouse HSF1 in samples. No significant cross-reactivity or interference between Mouse HSF1 and analogues was observed.
Mouse
1 Kit
3297
0.469 ng/mL
Storage is content dependent.
HSF1
Biological Fluids, Plasma, Serum