Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Mouse IgE ELISA Ready-SET-Go!™ Kit Non-distribution product as customer accommodation.

For quantitative detection of mouse IgE.

Manufacturer:  Invitrogen™ 885046022

 View more versions of this product

Catalog No. 50-112-8838


Add to cart

Includes: Capture Antibody Concentrate
Detection Antibody Concentrate
mouse IgE isotype control (standard), lyophilized, 500ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Pre-titrated Streptavidin-HRP
Tetramethylbenzidine (TMB) Substrate Solution
96 Well Plate included with product Cat. #’s ending in suffixes -22, -76, -86

Description

Description

IgE is one of the five classes of immunoglobulins found in mammals. Monomers consist of two heavy (epsilon) chains with 4 Ig-like constant domains, and two light chains. IgE plays an essential role in type I hypersensitivity (allergic responses such as hay fever, asthma, hives, anaphylatic shock) by binding to Fc receptors on basophils and mast cells. Crosslinking of IgE bound to Fc receptors induces degranulation of mast cells and basophils that promote allergic manifestations. IgE is also functionally important for immunity against parasites.
Specifications

Specifications

4-250 ng/mL
ELISA
Colorimetric Microplate Reader
ELISA Kit
RUO
Plasma,Serum,Supernatant
Plasma,10 μL|Serum,10 μL|Supernatant,50 μL
2-8°C
24 hr 30 min
Immunoglobulin E
Biotin
96-well plates
10X Coating Buffer: Buffer for plating the Capture Antibody
2 x 96 tests
4 ng/mL
50μL (prediluted 1:10)
Mouse
1 hr 45 min
Mouse
HRP
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only