Learn More
Mouse IgE Rat anti-Mouse, Alexa Fluor 488, Clone: LO-ME-3, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142334AF488
Catalog No. NBP142334AG
Add to cart
Description
Description
Mouse IgE Monoclonal specifically detects Mouse IgE in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
Mouse IgE
LO-ME-3
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
The antibody reacts with the Mouse IgE antigen.
Protein A or G purified
RUO
Primary
Mouse epsilon heavy chain of immunoglobulin (determined by immunodot).
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Rat
IgG1 Kappa
0.2 ml
Store at 4C in the dark.
Monoclonal
Mouse