Learn More
Mouse IgE Rat anti-Mouse, DyLight 488, Clone: LO-ME-3, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142334G
Catalog No. NBP142334G
Add to cart
Description
Description
Mouse IgE Monoclonal antibody specifically detects Antigen in Mouse samples. It is validated for Enzyme Linked Immunosorbent Assay, Immunohistochemistry (Frozen).
Specifications
Specifications
Mouse IgE
LO-ME-3
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
The antibody reacts with the Mouse IgE antigen.
Protein A or G purified
RUO
Primary
Mouse epsilon heavy chain of immunoglobulin (determined by immunodot).
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Rat
IgG1 Kappa
0.2 ml
Store at 4C in the dark.
Monoclonal
Mouse