Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IGFBP-6 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7761SP
This item is not returnable.
View return policy
Description
IGFBP-6 Monoclonal specifically detects IGFBP-6 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
IGFBP-6 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IBP6, IBP-6, IGF binding protein 6, IGF-binding protein 6, IGFBP6, IGFBP-6, insulin-like growth factor binding protein 6, insulin-like growth factor-binding protein 6 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3489 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, ELISA | |
122312 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
P47880 | |
IGFBP6 | |
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-6 Gly24-Gly238 Accession # P47880 | |
25 μg | |
Primary | |
Detects mouse IGFBP-6 in ELISAs and Western blots. In Western blots, 10% cross-reactivity with recombinant human (rh) IGFBP-3 and rhIGFBP-5 is observed and no cross-reactivity with rhIGFBP-1, recombinant mouse (rm) IGFBP-1, rhIGFBP-2, rmIGFBP-2, rmIGFBP-3, rhIGFBP-4, rmIGFBP-5, or rhIGFBP-6 is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction