Learn More
Invitrogen™ Mouse IgG Isotype Control
Description
Mouse IgG is purified by various methods from pooled normal mouse serum. The product is pure by SDS-PAGE analysis. The protein concentration is 2.5 mg/mL based on an E (1%) = 14.0 at 280 nm.
Specifications
Specifications
| Antigen | Mouse IgG |
| Applications | Control |
| Classification | Polyclonal |
| Concentration | 2.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene Alias | IgG1; Igh-4; Ighg1; Immunoglobulin G; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; VH7183 |
| Host Species | Mouse |
| Immunogen | Gamma Immunoglobin |
| Purification Method | Purified |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.