Learn More
Raybiotech IncSupplier Diversity Partner Mouse IgG1 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELMIGG11
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.8 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse IgA, IgE, IgM, IgG2a, IgG2b, IgG3, IgG4.
Catalog No. 50-195-0025