Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1, Fluorescein, Isotype control, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196849

Catalog No. NBP196849


Add to cart

Description

Description

IgG1 Monoclonal antibody specifically detects IgG1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications

Specifications

IgG1
Fluorescein
0.02 M Potassium Phosphate, 0.5 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
Mouse
Immunogen affinity purified
RUO
Primary
Reconstitute with 0.1 ml deionized water (or equivalent) to desired concentration.
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Western Blot 1:100 - 1:2000, Flow Cytometry 1:1000 - 1:5000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Conjugated
IgG1
0.1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Typically less than 1% cross reactivity against other mouse heavy or light chains isotypes was detected by ELISA.
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit