Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1 Isotype Control, Clone: 11711, R&D Systems™
GSA_VASDP

Catalog No. MAB002 Shop All Novus Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB002 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB002 Supplier R&D Systems Supplier No. MAB002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 193 publications

IgG1 Monoclonal specifically detects IgG1 in Mouse samples. It is validated for Control.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen IgG1
Applications Control
Classification Monoclonal
Clone 11711
Concentration LYOPH
Conjugate Unconjugated
Dilution Control
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Gene Symbols IGHG1
Host Species Mouse
Immunogen KLH
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Test Specificity Selected for use as mouse IgG 1 isotype control. In direct ELISAs, does not cross-react with recombinant human (rh) 6Ckine, recombinant mouse (rm) 6Ckine, rhACT-II, rhBLC/BCA-1, rmBLC, rmC10, recombinant rat (rr) CINC-1, rrCINC-2 alpha , rrCINC-2 beta , rrCINC-3, rhCK beta 8-1, recombinant viral (rv) CMV UL146, rmCRG-2, rhENA-78, rhEotaxin, rmEotaxin, rhEotaxin-2, rmEotaxin-2, rhEotaxin-3, rhFractalkine, rmFractalkine, rrFractalkine, rhGCP-2, rmGCP-2, rhGRO alpha , rhGRO beta , rhGRO gamma , rhHCC-1, rhI-309, rhI-Linck, rhIL-8, recombinant porcine IL-8, rhIP-10, rhI-TAC, rmJE, rmKC, rhLeukotactin-1, rhLymphotactin, rmLymphotactin, rmMARC, rhMCP-1, rhMCP-2, rhMCP-3, rhMCP-4, rmMCP-5, rvMCV type 2, rhMDC, rmMDC, rhMIG, rmMIG, rhMIP-1 alpha , rmMIP-1 alpha , rmMIP-1 beta , rhMIP-1 delta , rmMIP-1 gamma , rmMIP-2, rhMIP-3 alpha , rmMIP-3 alpha , rrMIP-3 alpha , rhMIP-3 beta , rmMIP-3 beta , rvMIP-I, rvMIP-II, rvMIP-III, rhMPIF-1, rhNAP-2, rhPARC, rhRANTES, rmRANTES, rhSDF-1 alpha , rmSDF-1 alpha , rhSDF-1 beta , rhTARC, rmTARC, rmTCA-3, rhTeck, or rmTeck.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG1 κ
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.