Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG1 Isotype Control, Novus Biologicals™

Mouse Polyclonal Antibody
Supplier: Novus Biologicals DDXCM01
Description
IgG1 Polyclonal specifically detects IgG1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular), Immunofluorescence.Specifications
| IgG1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-NaCl, pH 8. with No Preservative | |
| IGHG1 | |
| Ion exchange chromatography | |
| RUO | |
| Isotype Control (Primary) | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry, Immunofluorescence | |
| 0.5 mg/mL | |
| Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Flow (Cell Surface) 1:10 - 1:1000, Flow (Intracellular) 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500 | |
| IgG1, Igh-4, RGD1359539, VH7183 | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction