Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), Functional Grade, eBioscience™, Invitrogen™

Mouse Isotype Control

Supplier:  Invitrogen 16471482

Encompass_Preferred

 View more versions of this product

Catalog No. 50-145-33

  • $120.70 / Each of 1

    Save 15%

    Reg : $142.00

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Edge
Add to Cart

Description

Description

Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: This mouse IgG1 isotype control has been reported for use in surface and intracellular flow cytometric analysis, immunohistochemcial staining, immunoprecipitation and immunoblotting (WB). Applications Tested: This Mouse IgG1 isotype control has been tested by flow cytometric analysis of human peripheral leukocytes and can be used at the same concentration as the experimental antibody. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IgG1 kappa
P3.6.2.8.1
Functional Grade
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
Affinity chromatography
RUO
Not Applicable
Liquid
Control, Flow Cytometry, Functional Assay
1 mg/mL
PBS with no preservative; pH 7.2
Mouse
100 μg
Isotype Control (Primary)
4° C
IgG1 κ
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only.