Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG1 Kappa Light Chain, Clone: P3.6.2.8.1, FITC, Isotype control, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP1439560.05MG
Description
IgG1 Kappa Monoclonal specifically detects IgG1 Kappa in Mouse samples. It is validated for Flow Cytometry.Specifications
IgG1 Kappa Light Chain | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:100 - 1:2000 | |
Mouse | |
Primary | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
P3.6.2.8.1 | |
FITC | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide | |
0.05 mg | |
This monoclonal mouse IgG1, kappa immunoglobulin is useful as an isotype control. | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction