Learn More
Mouse IgG1 Mouse anti-Human, Alexa Fluor™ 594, Clone: 43D17, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222028AF594
Catalog No. NP222028A94
Add to cart
Description
Description
Mouse IgG1 Monoclonal antibody specifically detects Mouse IgG1 in Human samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
Mouse IgG1
43D17
Western Blot, ELISA
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant human IgG1 Fc domain. Mouse anti human IgG1 Fc domain.
0.1 mL
Primary
Human