Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1 Mouse anti-Human, Alexa Fluor™ 750, Clone: 43D17, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222028AF750

Catalog No. NP222028A70


Add to cart

Description

Description

Mouse IgG1 Monoclonal antibody specifically detects Mouse IgG1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Mouse IgG1
43D17
Western Blot, ELISA
Conjugated, Purified
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant human IgG1 Fc domain. Mouse anti human IgG1 Fc domain.
Protein G purified
Store at 4°C in the dark.
Monoclonal
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1
0.1 mL
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit