Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG1 Fc Mouse anti-Human, DyLight 550, Clone: 43D17, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222028R
Description
IgG1 Fc Monoclonal antibody specifically detects IgG1 Fc in Human samples. It is validated for Western Blot, ELISA.Specifications
IgG1 Fc | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
43D17 | |
Western Blot, ELISA | |
IgG1, Igh-4, RGD1359539, VH7183 | |
Human recombinant IgG1 Fc | |
0.1 mL | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title