Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG1, Texas Red, Isotype control, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP196991
Description
IgG1 Monoclonal specifically detects IgG1 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.Specifications
IgG1 | |
Texas Red | |
Lyophilized from 0.02 M Potassium Phosphate, 0.5 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
Mouse | |
0.1 mL | |
Primary | |
Reconstitute with 100 ul deionized water (or equivalent). | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Flow Cytometry 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:100 - 1:2000 | |
IgG1, Igh-4, RGD1359539, VH7183 | |
Protein A purified | |
RUO | |
Isotyping assay resulted the material is non-reactive with antisera to mouse IgG2a, IgG2b, IgG3, IgM, and IgA. Texas Red(TM) Conjugated Mouse IgG1 was proven by Dot Blot. | |
Mouse | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction