Learn More
Novus Biologicals™ Mouse IgG2a ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP262154
Catalog No. NBP262154
Add to cart
Description
Description
The Mouse IgG2a ELISA Kit (Colorimetric) measures mouse IgG2a in culture supernatants and serum.
Specifications
Specifications
1 Kit
7.81 - 250 ng/ml
ELISA
318.8 pg/ml
IgG2a
Culture Supernatant, Serum
Crossreactivity: Mouse IgG2a (100%), mouse IgG3 (0.49%), mouse IgM (0.12%) and <0.01%: rat IgG2a, mouse IgG1, mouse IgG2b
ELISA Kit (Colorimetric)
Storage of components varies. See protocol for specific instructions.
Mouse