Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG2a, HRP, Isotype control, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197075
Description
IgG2a Monoclonal specifically detects IgG2a in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.Specifications
IgG2a | |
HRP | |
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate | |
Protein A purified | |
RUO | |
Isotyping assay resulted the material is non-reactive with antisera to mouse IgG1, IgG2b, IgG3, IgM, and IgA. Peroxidase Conjugated Mouse IgG2a was proven by Dot Blot. | |
Mouse | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Flow Cytometry 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:100 - 1:2000 | |
Mouse | |
0.1 mg | |
Primary | |
Reconstitute with 100 ul deionized water (or equivalent). | |
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction