Learn More
Mouse IgG2a, clone: ZX4, Isotype Control, Invitrogen™
Mouse Isotype Control
Manufacturer:  Invitrogen SA112079
Catalog No. 01-675-858
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
SA1-12079 has been successfully used in FACS procedures to assess non-specific binding of mouse monoclonal antibodies in human samples.
Specifications
Specifications
Mouse IgG2a
Control
1mg/mL
PBS with 0.08% sodium azide
IgG2, IgG2a
White blood cells
Protein A/G
RUO
Isotype Control
Not Applicable
Control, Flow Cytometry
ZX4
Unconjugated
Liquid
Murine
IgG2a, κ
100μg
-20° C, Avoid Freeze/Thaw Cycles, Avoid Freeze/Thaw Cycles
Monoclonal