Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG2a Kappa Light Chain, Clone: eBM2a, Isotype control, Novus Biologicals™
Mouse Isotype Control
Supplier: Novus Biologicals NBP1433180.5MG
Specifications
IgG2a Kappa Light Chain | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
Mouse | |
0.5 mg | |
KLH and Freund's Adjuvant | |
Store at 4C in the dark. | |
IgG2a κ |
Flow Cytometry | |
M2a | |
Unconjugated | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide | |
KLH and Freund's Adjuvant | |
Primary | |
Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction