Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2a Kappa Light Chain, Clone: M2a, PerCP/Cy5.5, Isotype control, Novus Biologicals™

Mouse Isotype Control

Manufacturer:  Novus Biologicals NBP1428380.1MG

Catalog No. NBP14283801

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

IgG2a Kappa Light Chain
M2a
Flow Cytometry 1:10 - 1:1000
Conjugated
IgG2a Kappa
Store at 4C in the dark.
Monoclonal
Mouse
Flow Cytometry
PerCP-Cyanine5.5
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Mouse
0.1 mg
Primary
This monoclonal mouse IgG2a, kappa immunoglobulin is used as an isotype control for mouse IgG2a antibodies.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit