Learn More
Crystal Chem IncSupplier Diversity Partner Mouse IgG2B ELISA Kit, 96-wells DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80648
The Mouse IgG2B ELISA is used to quantify levels of mouse IgG2B in serum and plasma using only a 5 µL sample volume. Highlights: Kit uses sample size of only 5 µL. High sensitivity of 0.88 ng/mL. Dynamic range of 6.25 - 200 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7778