Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Mouse IgG2b ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals™ NBP262155
Description
The Mouse IgG2b ELISA Kit (Colorimetric) measures mouse IgG2b in culture supernatants and serum.Specifications
7.81 - 250 ng/ml | |
Crossreactivity: Mouse IgG2b (100%) and <0.10%: rat IgG2b, mouse IgG1, mouse IgG2a, mouse IgG3, mouse IgM | |
Mouse | |
1 Kit | |
Store at 4°C |
1.05 ng/ml | |
ELISA | |
ELISA Kit (Colorimetric) | |
Culture Supernatant,Serum | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title