Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2b kappa Isotype Control (eBMG2b), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Mouse Isotype Control

Supplier:  Invitrogen 53473280

Encompass_Preferred

Catalog No. 50-167-51


Only null left
Edge
Add to Cart

Description

Description

Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin. Applications Reported: Alexa Fluor® 488 Mouse IgG2b Isotype Control has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis. Applications Tested: Alexa Fluor® 488 Mouse IgG2b Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells. Use isotype control at the same concentration as experimental antibody. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

Mouse IgG2b is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the third most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2b is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2b is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IgG2b kappa
eBMG2b
Alexa Fluor 488
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Affinity chromatography
RUO
Not Applicable
Liquid
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Mouse
25 μg
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only.