Learn More
Invitrogen™ Mouse IgG2b kappa Isotype Control (eBMG2b), PE-Cyanine7, eBioscience™
Mouse Monoclonal Isotype Control
Supplier: Invitrogen™ 25473281
Description
Description: The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin. Applications Reported: PE-Cy7 Mouse IgG2b Iso Cntrl has been reported for use in flow cytometric analysis. Applications Tested: Mouse IgG2b K Isotype Control PE-Cyanine7 has been tested by flow cytometric analysis of normal human peripheral blood cells. Use isotype control at the same concentration as experimental antibody. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-822-49) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-54) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2b is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the third most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2b is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2b is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
Specifications
| Mouse IgG2b kappa | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Mouse | |
| 50 μg | |
| Isotype Control (Primary) | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Control, Flow Cytometry | |
| eBMG2b | |
| PE-Cyanine7 | |
| IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
| Affinity chromatography | |
| RUO | |
| Not Applicable | |
| Isotype Control | |
| IgG2b κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.