Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2c Rabbit anti-Mouse, Clone: RM223, Abnova™

Rabbit Monoclonal Antibody

$379.00

Specifications

Antigen Mouse IgG2c
Clone RM223
Host Species Rabbit
Isotype IgG
Target Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-035-318 View Documents Abnova Corporation
MAB15110
100 ug Each for $379.00
Add to cart
 
89-035-350 View Documents Abnova Corporation
MAB15161
100 ug Each for $379.00
Add to cart
 
Description

Description

Rabbit monoclonal antibody raised against native mouse IgG2c.

  • High specificity
  • High reactivity
Specifications

Specifications

Mouse IgG2c
Rabbit
Mouse
RUO
Liquid
Primary
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
RM223
IgG
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit