Learn More
Mouse IgG3, Fluorescein, Isotype control, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196847
Catalog No. NBP196847
Add to cart
Description
Description
IgG3 Monoclonal antibody specifically detects IgG3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Dot Blot, Fluorophore-linked immunosorbent assay.
Specifications
Specifications
IgG3
Fluorescein
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809
Mouse
Protein A purified
RUO
Primary
Typically less than 1% cross reactivity against other mouse heavy or light chain specific antibodies was detected by ELISA.
Mouse
Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Western Blot 1:100 - 1:2000, Flow Cytometry 1:1000 - 1:5000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Dot Blot 1:100 - 1:2000, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Conjugated
IGHG3
IgG3
0.1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
3502
Reconstitute with 0.1 ml deionized water (or equivalent) to desired concentration.