Learn More
Mouse IgG3, HRP, Isotype control, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197077
Catalog No. NBP197077
Add to cart
Description
Description
IgG3 Monoclonal antibody specifically detects IgG3 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
Specifications
IgG3
HRP
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Gentamicin Sulfate
DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809
Mouse
Protein A purified
RUO
Primary
Typically, less than 1% cross reactivity against other mouse and human heavy or light chains isotypes was detected by ELISA.
Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Flow Cytometry 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Conjugated
IGHG3
IgG3
0.1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
3502
Reconstitute with 0.1 ml deionized water (or equivalent) to desired concentration.