Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG3, Rhodamine, Isotype control, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196850

Catalog No. NBP196850


Add to cart

Description

Description

IgG3 Monoclonal antibody specifically detects IgG3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications

Specifications

IgG3
Rhodamine
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809
Mouse
Ion exchange chromatography
RUO
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse IgG3 and anti-Mouse Serum.
Mouse
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Western Blot 1:100 - 1:2000, Flow Cytometry 1:1000 - 1:5000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Conjugated
IGHG3
IgG3
0.1 ml
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
3502
Reconstitute with 0.1 ml deionized water (or equivalent) to desired concentration.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit