Learn More
Invitrogen™ Mouse IgM Isotype Control (11E10), APC, eBioscience™
Mouse Monoclonal Isotype Control
Supplier: Invitrogen™ 17475280
Description
Description: The mouse IgM is useful as an isotype control immunoglobulin. The immunoglobulin was generated by immunization with lipopolysaccharide (LPS). The immunoglobulin is reactive with LPS. Applications Reported: Mouse IgM Isotype Control APC has been reported for use in flow cytometric analysis. Applications Tested: Mouse IgM Isotype Control APC has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood cells. Use at the same concentration as the experimental antibody. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgM is an immunoglobulin M antibody that is produced by B cells in response to an antigen. It is the first antibody produced during an immune response and is the largest immunoglobulin in the serum. Mouse IgM is composed of five heavy chains and five light chains, and it is involved in complement activation and agglutination of antigens. It plays a crucial role in the innate immune response and is important for neutralizing pathogens before the adaptive immune response is fully activated. Mouse IgM is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry.
Specifications
| Mouse IgM | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Mouse | |
| 25 μg | |
| Isotype Control (Primary) | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Control, Flow Cytometry | |
| 11E10 | |
| APC | |
| AGM1; AI326478; Igh6; Igh-6; Ighm; Igh-M; Igm; immunoglobulin heavy constant mu; MU; muH; VH | |
| Affinity chromatography | |
| RUO | |
| Not Applicable | |
| Isotype Control | |
| IgM |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.