Learn More
Mouse IgM Isotype Control, Invitrogen™
Mouse Isotype Control
Supplier: Invitrogen 026800

Description
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
This antibody will react with sheep red blood cells, but is suitable as a negative reagent control for most applications.
Specifications
Mouse IgM | |
1mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Mouse | |
Purified | |
RUO | |
None | |
Liquid |
ELISA | |
Unconjugated | |
IgM, IgM mu | |
Mouse IgM | |
1 mg | |
Isotype Control (Primary) | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
IgM |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.