Learn More
Mouse IgM Rat anti-Mouse, DyLight 405, Clone: LO-MM-9, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142328V
Catalog No. NBP142328V
Add to cart
Description
Description
Mouse IgM Monoclonal antibody specifically detects Antigen in Mouse samples. It is validated for Enzyme Linked Immunosorbent Assay, Immunohistochemistry (Frozen).
Specifications
Specifications
Mouse IgM
LO-MM-9
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
IGHM
The antibody reacts with the Mouse IgM antigen.
Protein A or G purified
RUO
Primary
Mouse mu heavy chain of immunoglobulin (determined by immunodot). Avidity on IgM: 7 * 108 M-1. No cross reactivity with other animals.
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH
Rat
IgG2a Kappa
0.2 ml
Store at 4C in the dark.
Monoclonal
Mouse