Learn More
Mouse IgM Rat anti-Mouse, DyLight 550, Clone: LO-MM-9, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142328R
Catalog No. NBP142328R
Add to cart
Description
Description
IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
Mouse IgM
LO-MM-9
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
IGHM
The antibody reacts with the Mouse IgM antigen.
Protein A or G purified
RUO
Primary
Mouse mu heavy chain of immunoglobulin (determined by immunodot). Avidity on IgM: 7 * 10^8 M-1. No cross reactivity with other species.
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH
Rat
IgG2a Kappa
0.2 ml
Store at 4C in the dark.
Monoclonal
Mouse