Learn More
Novus Biologicals™ Mouse IL-1 alpha/IL-1F1 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192673
Catalog No. NBP192673
Add to cart
Description
Description
The Mouse IL-1 alpha/IL-1F1 ELISA Kit (Colorimetric) measures mouse IL-1 alpha/IL-1F1 in cell culture supernatant, serum.
Specifications
Specifications
7.8 - 500pg/mL
3552
ELISA Kit
4.0pg/mL
Mouse (Mu)
IL-1 alpha/IL-1F1
IL1A
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-1alpha, Biotin-Conjugate anti-mouse IL-1alpha monoclonal antibody, Streptavidin-HRP, Mouse IL-1alpha Standard lyophilized, 1000 pg/ml upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
Apoptosis, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Hematopoietin-1, IL1, IL-1 alpha, IL-1A, IL1-ALPHA, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha