Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-1 beta ELISA Standard,Invitrogen™ Non-distribution product as customer accommodation.

Recombinant Protein

Manufacturer:  Invitrogen 29801265

Catalog No. 50-112-3485


Add to cart

Alert:

Contains Sodium azide


Description

Description

Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value. Description: Mouse IL-1 beta, also called lymphocyte activating factor (LAF), endogenous pyrogen (EP), leucocyte endogenous mediator (LEM), mononuclear cell factor (MCF), is a ~17 kDa factor produced by a wide variety of cells, including macrophages, dendritic cells, T and B cells. IL-1 beta is mostly cell associated with 23% amino acid homology with IL-1 alpha. The immune regulatory role of IL-1 beta is exerted on a wide range of cells including lymphocytes, epithelial cells and fibroblasts. In vivo, it induces hypotension, fever, and acute phase response. Applications Reported: Mouse IL-1 beta RSG ELISA Lyophilized Standard has been reported for use in the Mouse IL-1 beta ELISA RSG (cat. 88-7013). Applications Tested: The intended use of the Mouse IL-1 beta RSG ELISA Lyophilized Standard is as a standard for Mouse IL-1 beta ELISA (cat. 88-7013). Typically the standard is run in a series of 7 doubling dilutions from the  ft.top ft. standard. Please refer to the Certificate of Analysis included with each ELISA set for instructions on the proper reconstitution and dilution of the lyophilized standard. Note: These are single-use vials. Use one time within 4 hours of reconstitution and discard. Reconstitute and dilute as suggested in the Certificate of Analysis or recovery may not be accurate. Source: E. coli expressed amino acids Val 118-Ser 269 of mouse IL-1 beta accession &Num; P10749.
Specifications

Specifications

P10749
Lyophilized
16176
Affinity chromatography
E. coli-expressed amino acids Val 118-Ser 269 of mouse IL-1 beta (accession # P10749).
RUO
Mouse IL-1 beta ELISA Standard
Control, ELISA
Unlabeled
E. coli
Control, ELISA
protein with no preservative
Mouse IL-1 beta ELISA Standard
4pack
4°C
Recombinant
Il1b
Mouse
Ms
Recombinant
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit