Learn More
R&D Systems™ Mouse IL-10 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
Mouse IL-10 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  R&D Systems™ M1000B
 View more versions of this product
Catalog No. M1000B
Add to cart
This item is not returnable. View return policy
Description
Description
The Mouse IL-10 Quantikine ELISA Kit quantifies mouse IL-10 in cell culture supernates (25 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
15.60 - 1000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
16153
5.22pg/mL
Natural and recombinant mouse IL-10
Sufficient materials to run ELISAs on at least two 96-well plates
1 Kit
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
HRP
ELISA
Solid Phase Sandwich ELISA
Cell Culture Supernates (25uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Mouse
2 Plates