Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Mouse IL-10 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
$89.50 - $5760.00
Specifications
Accession Number | P18893 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 16153 |
Molecular Weight (g/mol) | 18.7 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
21010100UG
|
Gibco™
21010100UG |
100 μg |
Each for $1,154.00
|
|
|||||
210-10-10UG
|
Gibco™
2101010UG |
10 μg |
Each for $217.00
|
|
|||||
210-10-1MG
|
Gibco™
210101MG |
1 mg |
Each for $5,760.00
|
|
|||||
21010250UG
|
Gibco™
21010250UG |
250 μg |
Each for $2,015.00
|
|
|||||
210-10-2UG
|
Gibco™
210102UG |
2 μg |
Each for $89.50
|
|
|||||
21010500UG
|
Gibco™
21010500UG |
500 μg |
Each for $3,460.00
|
|
|||||
210-10-50UG
|
Gibco™
2101050UG |
50 μg |
Each for $692.00
|
|
|||||
Description
Interleukin 10 (IL-10, CSIF) is an anti-inflammatory cytokine mainly produced by macrophages and Th2 cells. IL-10 is an 18.6 kDa protein of 160 amino acid residues that shares over 80% sequence homology with the Epstein-Barr Virus protein (BCRFI). The reported biological activities of IL-10, which maybe interrelated, include inhibition of macrophage-mediated cytokine synthesis, suppression of the delayed-type hyper-sensitivity response, and stimulation of the Th2 cell response which results in elevated antibody production. IL-10 functions by inhibiting pro-inflammatory cytokines made by macrophages and regulatory T cells including IFN-gamma, TNF-alpha, IL-2, and IL-3, IL-4, and GM-CSF. IL-10 is also known to suppress antigen presentation on antigen presenting cells, enhances B cell survival, proliferation, and antibody production. IL-10 can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. While human IL-10 can act on murine cells, murine IL-10 cannot act on human cells. Binds to a retinoid X receptor (RXR) responsive element from the cellular retinol-binding protein II promoter (CRBPII-RXRE). IL-10 inhibits the 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element. Knockout studies in mice suggest the function of this cytokine as an essential immunoregulator in the intestinal tract.Specifications
P18893 | |
protein with no preservative | |
18.7 | |
E. coli | |
RUO | |
CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF | |
Mouse | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
16153 | |
Mouse IL-10 | |
-20°C | |
<1 EU/μg | |
IL10 | |
Unconjugated | |
MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title