Learn More
Novus Biologicals™ Mouse IL-12 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192677
Catalog No. NBP192677
Add to cart
Description
Description
The Mouse IL-12 ELISA Kit (Colorimetric) measures mouse IL-12 in cell culture supernatant, serum.
Specifications
Specifications
31.3 - 2,000pg/mL
3592
ELISA Kit
4.0pg/mL
Mouse (Mu)
IL-12
IL12A
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-12, Biotin-Conjugate anti-mouse IL-12 monoclonal antibody, Streptavidin-HRP, Mouse IL-12 Standard lyophilized, 4ng/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
Immunology, Innate Immunity
CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha cha