Learn More
Novus Biologicals™ Mouse IL-13 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192663
Catalog No. NBP192663
Add to cart
Description
Description
The Mouse IL-13 ELISA Kit (Colorimetric) measures mouse IL-13 in cell culture supernatant, serum, plasma (edta, citrate).
Specifications
Specifications
7.8 - 500pg/mL
3596
ELISA Kit
2.8pg/mL
Mouse (Mu)
IL-13
IL13
ELISA
Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to mouse IL-13, Biotin-Conjugate anti-mouse IL-13 polyclonal antibody, Streptavidin-HRP, Mouse IL-13 Standard lyophilized, 1ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, citrate)
Storage of components varies. See protocol for specific instructions.
Asthma, Cancer, Cytokine Research, Immunology
ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600