Learn More
Novus Biologicals™ Mouse IL-17F ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192666
Catalog No. NBP192666
Add to cart
Description
Description
The Mouse IL-17F ELISA Kit (Colorimetric) measures mouse IL-17F in cell culture supernatant, serum and plasma (edta).
Specifications
Specifications
62.5 - 4,000pg/mL
112744
ELISA Kit
10pg/mL
Mouse (Mu)
IL-17F
IL17F
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-17F, Biotin-Conjugate anti-mouse IL-17F polyclonal antibody, Streptavidin-HRP, Mouse IL-17F Standard lyophilized, 4ng/mL upon reconstitution, Sample Diluent, Conjugate Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Calibrator Diluent, Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum and plasma (EDTA)
Storage of components varies. See protocol for specific instructions.
Cytokine Research
Cytokine ML-1, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24