Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-21 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 1 publication
$245.00 - $479.50
Specifications
Antigen | IL-21 |
---|---|
Clone | 149204 |
Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
IL-21 Monoclonal specifically detects IL-21 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-21 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CVID11, IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11 | |
IL21 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse IL-21 in Western blots. Does not cross-react with recombinant human IL-21, recombinant mouse (rm) IL-15, or rmIL-12. |
149204 | |
Flow Cytometry | |
APC | |
Rat | |
Mouse | |
Q9ES17 | |
59067 | |
E. coli-derived recombinant mouse IL-21 Pro25-Ser146 Accession # Q9ES17 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title