Learn More
Raybiotech IncSupplier Diversity Partner Mouse IL-4 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELMIL4CL1
For cell and tissue lysate samples. Sensitivity: 1 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes mouse IL-4.
Catalog No. 50-195-0058