Learn More
Novus Biologicals™ Mouse IL-4 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192675B
Catalog No. NBP192675B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The mouse IL-4 ELISA is an enzyme-linked immunosorbent assay for the quantitative detection of mouse IL-4.
Specifications
Specifications
3.9 - 250pg/mL
3565
ELISA Kit
2.0pg/mL
Mouse (Mu)
IL-4
IL4
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-4, Biotin-Conjugate anti-mouse IL-4 monoclonal antibody, Streptavidin-HRP, Mouse IL-4 Standard lyophilized, 500 pg/ml upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Adhesive Films, Stop Solution (1M Phosphoric acid)
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra