Learn More
Novus Biologicals™ Mouse IL-5 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192672
Catalog No. NBP192672
Add to cart
Description
Description
The Mouse IL-5 ELISA Kit (Colorimetric) measures mouse IL-5 in cell culture supernatant, serum.
Specifications
Specifications
7.8 - 500pg/mL
3567
ELISA Kit
3.3pg/mL
Mouse (Mu)
IL-5
IL5
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-5, Biotin-Conjugate anti-mouse IL-5 monoclonal antibody, Streptavidin-HRP, Mouse IL-5 Standard lyophilized, 1000 pg/ml upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology
B-cell differentiation factor I, EDF, Eosinophil differentiation factor, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRFB cell differentiation factor I